missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-phenylphenol 96.0+%, TCI America™

Catalog No. C026325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C026325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C026325G Supplier TCI America Supplier No. C026325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92-04-6
Molecular Formula C12H9ClO
Molecular Weight (g/mol) 204.653
MDL Number MFCD00045745
InChI Key BZWMYDJJDBFAPE-UHFFFAOYSA-N
Synonym sanidril, 4-phenyl-2-chlorophenol, 3-chloro-4-hydroxybiphenyl, dowicide 4, 3-chlorobiphenyl-4-ol, 3-chloro-4-biphenylol, o-chloro-p-phenylphenol, 2-chloro-p-phenylphenol, 3-chloro-4-hydroxydiphenyl, phenol, 2-chloro-4-phenyl
PubChem CID 7074
IUPAC Name 2-chloro-4-phenylphenol
SMILES C1=CC=C(C=C1)C2=CC(=C(C=C2)O)Cl

Specifications

Melting Point 79°C
Color Red-White
Boiling Point 322°C
Quantity 25 g
Formula Weight 204.65
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-phenylphenol
RTECSNumber DV6650000
TSCA Yes