accessibility menu, dialog, popup

UserName

OHCl

CAS RN 92-04-6

IUPAC Name: 3-chloro-[1,1'-biphenyl]-4-ol
Synonyms: 4-phenyl-2-chlorophenol
Molecular Weight (g/mol): 204.65
Molecular Formula: C12H9ClO
InChi Key: BZWMYDJJDBFAPE-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1Cl)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results