missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-nitrobenzamide, 98%

Catalog No. p-7024654
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1970206 5 g
AAA1970214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1970206 Supplier Thermo Scientific Chemicals Supplier No. A1970206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3011-89-0 | C7H5ClN2O3 | 200.578 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3011-89-0
Molecular Formula C7H5ClN2O3
Molecular Weight (g/mol) 200.578
MDL Number MFCD00017119
InChI Key GFGSZUNNBQXGMK-UHFFFAOYSA-N
Synonym aklomide, aklomix, clomide, alkomide, benzamide, 2-chloro-4-nitro, component of aklomix, component of novastat-w, unii-b0e341ra20, 2-chloro-4-nitro benzamide, dsstox_cid_21405
PubChem CID 2075
IUPAC Name 2-chloro-4-nitrobenzamide
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)C(=O)N

Specifications

Melting Point 168°C to 172°C
Quantity 5 g
Beilstein 1966537
Formula Weight 200.58
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-nitrobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 221-143-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only