missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methylaniline 98.0+%, TCI America™

Catalog No. a143725g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
A143725G 25 g
A14375G 5 g
2 options

Catalog No. A143725G

Supplier: TCI America A143725G

Only null left

Chemical Identifiers

CAS 615-65-6
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00007666
InChI Key XGYLSRFSXKAYCR-UHFFFAOYSA-N
Synonym 2-chloro-p-toluidine, benzenamine, 2-chloro-4-methyl, 4-amino-3-chlorotoluene, p-toluidine, 2-chloro, 2-chlor-4-toluidin, 4-methyl-2-chloroaniline, unii-k59isj2ko1, 2-chlor-4-toluidin czech, ccris 2887, 3-chloro-4-aminotoluene
PubChem CID 12007
IUPAC Name 2-chloro-4-methylaniline
SMILES CC1=CC(=C(C=C1)N)Cl

Specifications

Melting Point 7°C
Boiling Point 224°C
Quantity 25 g
UN Number 3429
Formula Weight 141.60
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-methylaniline
EINECSNumber (3)-0265
RTECSNumber XU5110000
TSCA Yes