missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-methoxytoluene 95.0+%, TCI America™
Supplier: TCI America C319025G
Specifications
| 2-Chloro-4-methoxytoluene | |
| Yellow | |
| C8H9ClO | |
| 25 g | |
| ZMZVVVASCILFJL-UHFFFAOYSA-N | |
| 2-chloro-4-methoxy-1-methylbenzene | |
| 6951757 | |
| ≥95.0% (GC) |
| 54788-38-4 | |
| 213°C | |
| MFCD00087376 | |
| 3-chloro-4-methylanisole, 2-chloro-4-methoxytoluene, 2-chloro-4-methoxy-1-methyl-benzene, benzene, 2-chloro-4-methoxy-1-methyl, pubchem4101, acmc-209xb6, ksc602k7p, benzene,2-chloro-4-methoxy-1-methyl | |
| CC1=C(C=C(C=C1)OC)Cl | |
| 156.609 | |
| 156.61 | |
| Liquid |
Chemical Identifiers
| 54788-38-4 | |
| 156.609 | |
| ZMZVVVASCILFJL-UHFFFAOYSA-N | |
| 6951757 | |
| CC1=C(C=C(C=C1)OC)Cl |
| C8H9ClO | |
| MFCD00087376 | |
| 3-chloro-4-methylanisole, 2-chloro-4-methoxytoluene, 2-chloro-4-methoxy-1-methyl-benzene, benzene, 2-chloro-4-methoxy-1-methyl, pubchem4101, acmc-209xb6, ksc602k7p, benzene,2-chloro-4-methoxy-1-methyl | |
| 2-chloro-4-methoxy-1-methylbenzene |
Safety and Handling
TSCA : No