accessibility menu, dialog, popup

UserName

CH3OCH3Cl

CAS RN 54788-38-4

IUPAC Name: 2-chloro-4-methoxy-1-methylbenzene
Synonyms: 2-chloro-4-methoxy-1-methylbenzene
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: ZMZVVVASCILFJL-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results