missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxytoluene 95.0+%, TCI America™

Catalog No. C31905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31905G 5 g
C319025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31905G Supplier TCI America Supplier No. C31905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 54788-38-4
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00087376
InChI Key ZMZVVVASCILFJL-UHFFFAOYSA-N
Synonym 3-chloro-4-methylanisole, 2-chloro-4-methoxytoluene, 2-chloro-4-methoxy-1-methyl-benzene, benzene, 2-chloro-4-methoxy-1-methyl, pubchem4101, acmc-209xb6, ksc602k7p, benzene,2-chloro-4-methoxy-1-methyl
PubChem CID 6951757
IUPAC Name 2-chloro-4-methoxy-1-methylbenzene
SMILES CC1=C(C=C(C=C1)OC)Cl

Specifications

Color Yellow
Boiling Point 213°C
Quantity 5 g
Formula Weight 156.61
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-methoxytoluene