missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. C30775G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30775G 5 g
C30771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30775G Supplier TCI America Supplier No. C30775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21971-21-1
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00085943
InChI Key IBANGHTVBPZCHF-UHFFFAOYSA-N
Synonym 2-chloro-4-methoxy-benzoic acid, 2-chloro-4-methyloxy benzoic acid, benzoic acid, 2-chloro-4-methoxy, 2-chloro-4-methoxybenzoicacid, 2-chloranissaure, pubchem14030, 2-chloro-p-anisic acid, ksc494q0b, 4-methoxy-2-chloro-benzoic acid
PubChem CID 12648026
IUPAC Name 2-chloro-4-methoxybenzoic acid
SMILES COC1=CC(=C(C=C1)C(=O)O)Cl

Specifications

Melting Point 212°C
Quantity 5 g
Formula Weight 186.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-methoxybenzoic Acid