accessibility menu, dialog, popup

UserName

CAS RN 21971-21-1

IUPAC Name: 2-chloro-4-methoxybenzoic acid
Synonyms: 2-chloro-4-methoxybenzoic acid
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: IBANGHTVBPZCHF-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=C(C=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)