missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorotoluene 98.0+%, TCI America™

Catalog No. C122725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C122725G 25 g
C1227500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C122725G Supplier TCI America Supplier No. C122725G

Chemical Identifiers

CAS 452-73-3
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.573
MDL Number MFCD00000572
InChI Key CSARJIQZOSVYHA-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorotoluene, benzene, 2-chloro-4-fluoro-1-methyl, 4-fluoro-2-chlorotoluene, 2-chlor-4-fluor-1-methylbenzol, pubchem1605, 2-chloro-4fluorotoluene, acmc-1adyb, 4-fluor-2-chlor-toluol, ksc235q0r, csarjiqzosvyha-uhfffaoysa
PubChem CID 96747
IUPAC Name 2-chloro-4-fluoro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)F)Cl

Specifications

Color Yellow
Boiling Point 153°C
Quantity 25 g
UN Number 1993
Formula Weight 144.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-4-fluorotoluene