accessibility menu, dialog, popup

UserName

CH3FCl

CAS RN 452-73-3

IUPAC Name: 2-chloro-4-fluoro-1-methylbenzene
Synonyms: 2-chloro-4-fluoro-1-methylbenzene
Molecular Weight (g/mol): 144.57
Molecular Formula: C7H6ClF
InChi Key: CSARJIQZOSVYHA-UHFFFAOYSA-N
SMILES: CC1=CC=C(F)C=C1Cl
Boiling Point 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)