missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorothiophenol, 97%

Catalog No. AAA1349803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1349803 1 g
AAA1349806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1349803 Supplier Thermo Scientific Chemicals Supplier No. A1349803

CAS: 175277-99-3 | C6H4ClFS | 162.606 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 175277-99-3
Molecular Formula C6H4ClFS
Molecular Weight (g/mol) 162.606
MDL Number MFCD00052936
InChI Key KUAPPJSILOMQPC-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorothiophenol, 2-chloro-4-fluorobenzene-1-thiol, benzenethiol,2-chloro-4-fluoro, 2-chloro-4-fluoro-benzenethiol, benzenethiol, 2-chloro-4-fluoro, pubchem2850, acmc-1caad, 2-chloro-4-fluoro-thiophenol, 4-fluoro-2-chloro-benzenethiol, #
PubChem CID 519446
IUPAC Name 2-chloro-4-fluorobenzenethiol
SMILES C1=CC(=C(C=C1F)Cl)S

Specifications

Boiling Point 86°C to 88°C (15 mmHg)
Refractive Index 1.567
Quantity 1 g
Beilstein 8478226
Sensitivity Air Sensitive
Formula Weight 162.61
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorothiophenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H332-H315-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only