accessibility menu, dialog, popup

UserName

CAS RN 175277-99-3

IUPAC Name: (2-chloro-4-fluorophenyl)sulfanide
Synonyms: (2-chloro-4-fluorophenyl)sulfanide
Molecular Weight (g/mol): 161.6
Molecular Formula: C6H3ClFS
InChi Key: KUAPPJSILOMQPC-UHFFFAOYSA-M
SMILES: FC1=CC=C([S-])C(Cl)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)