missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$178.95 - $178.95
Specifications
| CAS | 313545-72-1 |
|---|---|
| Melting Point | 237°C |
| Color | White |
| Molecular Formula | C6H5BClFO2 |
| MDL Number | MFCD02684295 |
Specifications
| 313545-72-1 | |
| White | |
| MFCD02684295 | |
| (2-chloro-4-fluorophenyl)boronic acid | |
| 174.36 |
| 237°C | |
| C6H5BClFO2 | |
| XOFNMNLYGPKKOV-UHFFFAOYSA-N | |
| 17750229 | |
| Crystalline Powder |