accessibility menu, dialog, popup

UserName

HOBOHFCl

CAS RN 313545-72-1

IUPAC Name: (2-chloro-4-fluorophenyl)boronic acid
Synonyms: 2-chloro-4-fluorophenylboronic acid
Molecular Weight (g/mol): 174.36
Molecular Formula: C6H5BClFO2
InChi Key: XOFNMNLYGPKKOV-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(F)C=C1Cl
Quantity 
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)