missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorophenylacetonitrile, 97%

Catalog No. AAB2313206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2313206 5 g
AAB2313214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2313206 Supplier Thermo Scientific Chemicals Supplier No. B2313206
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 75279-56-0 | C8H5ClFN | 169.583 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75279-56-0
Molecular Formula C8H5ClFN
Molecular Weight (g/mol) 169.583
MDL Number MFCD00236029
InChI Key GSMCLMKFBYLWRP-UHFFFAOYSA-N
Synonym 2-chloro-4-fluorophenylacetonitrile, 2-chloro-4-fluorobenzyl cyanide, 2-2-chloro-4-fluorophenyl acetonitrile, 2-chloro-4-fluorobenzylcyanide, benzeneacetonitrile, 2-chloro-4-fluoro, 2-chloro-fluorophenylacetonitrile, 2-chloro-4-fluorophenyl acetonitrile, 2-2-chloro-4-fluorophenyl ethanenitrile, pubchem4251
PubChem CID 2725063
IUPAC Name 2-(2-chloro-4-fluorophenyl)acetonitrile
SMILES C1=CC(=C(C=C1F)Cl)CC#N

Specifications

Melting Point 67°C to 69°C
Quantity 5 g
UN Number UN3439
Formula Weight 169.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorophenylacetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H300-H311-H331-H315-H319
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only