accessibility menu, dialog, popup

UserName

CAS RN 75279-56-0

IUPAC Name: 2-(2-chloro-4-fluorophenyl)acetonitrile
Synonyms: 2-(2-chloro-4-fluorophenyl)acetonitrile
Molecular Weight (g/mol): 169.58
Molecular Formula: C8H5ClFN
InChi Key: GSMCLMKFBYLWRP-UHFFFAOYSA-N
SMILES: FC1=CC=C(CC#N)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results