missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoronitrobenzene 98.0+%, TCI America™

Catalog No. p-7125559
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C26155G 5 g
C261525G 25 g
2 options

Catalog No. C26155G

Supplier: TCI America C26155G

Only null left

Chemical Identifiers

CAS 2106-50-5
Molecular Formula C6H3ClFNO2
Molecular Weight (g/mol) 175.543
MDL Number MFCD03412200
InChI Key KQOOFMWRLDRDAX-UHFFFAOYSA-N
Synonym 2-chloro-4-fluoronitrobenzene, 1-chloro-5-fluoro-2-nitrobenzene, benzene, 2-chloro-4-fluoro-1-nitro, pubchem4342, acmc-209fh8, 2-choro-4-fluoronitrobenzene, ksc205m1p, 3-chloro-4-nitrofluorobenzene, 4-fluoro-2-chloronitrobenzene, 2-chloro-4-fluoronitrobenzenen
PubChem CID 75017
IUPAC Name 2-chloro-4-fluoro-1-nitrobenzene
SMILES C1=CC(=C(C=C1F)Cl)[N+](=O)[O-]

Specifications

Melting Point 37°C
Color Yellow
Boiling Point 71°C
Quantity 5 g
Formula Weight 175.54
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-fluoronitrobenzene