accessibility menu, dialog, popup

UserName

ONOFCl

CAS RN 2106-50-5

IUPAC Name: 2-chloro-4-fluoro-1-nitrobenzene
Synonyms: 2-chloro-4-fluoro-1-nitrobenzene
Molecular Weight (g/mol): 175.54
Molecular Formula: C6H3ClFNO2
InChi Key: KQOOFMWRLDRDAX-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(F)C=C1Cl
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)