missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzyl Alcohol 98.0+%, TCI America™

Catalog No. p-7125510
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25275G 5 g
C252725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25275G Supplier TCI America Supplier No. C25275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 208186-84-9
Molecular Formula C7H6ClFO
Molecular Weight (g/mol) 160.572
MDL Number MFCD00236024
InChI Key ZUHMDLLAHZUDRE-UHFFFAOYSA-N
PubChem CID 4982711
IUPAC Name (2-chloro-4-fluorophenyl)methanol
SMILES C1=CC(=C(C=C1F)Cl)CO

Specifications

Melting Point 47°C
Quantity 5 g
Formula Weight 160.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-fluorobenzyl Alcohol