accessibility menu, dialog, popup

UserName

CAS RN 208186-84-9

IUPAC Name: (2-chloro-4-fluorophenyl)methanol
Synonyms: (2-chloro-4-fluorophenyl)methanol
Molecular Weight (g/mol): 160.57
Molecular Formula: C7H6ClFO
InChi Key: ZUHMDLLAHZUDRE-UHFFFAOYSA-N
SMILES: OCC1=CC=C(F)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results