missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-fluorobenzyl Alcohol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 47°C |
| Quantity | 25 g |
| Formula Weight | 160.57 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Chloro-4-fluorobenzyl Alcohol |
Safety and Handling
| TSCA | No |