missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluorobenzoic acid, 98%

CAS: 2252-51-9 | C7H4ClFO2 | 174.555 g/mol

$48.19 - $155.70

Chemical Identifiers

CAS 2252-51-9
Molecular Formula C7H4ClFO2
Molecular Weight (g/mol) 174.555
MDL Number MFCD00010615
InChI Key GRPWQLDSGNZEQE-UHFFFAOYSA-N
Synonym benzoic acid, 2-chloro-4-fluoro, 2-chloro-4-fluoro-benzoic acid, 2-chloro-4-fluor obenzoic acid, pubchem1370, acmc-209fx5, ksc207e4n, rarechem al bo 0405, 2-chloro 4-fluoro benzoic acid, 2-chloro-4-fluoro benzoic acid, 4-fluoro-2-chloro-benzoic acid
PubChem CID 75259
IUPAC Name 2-chloro-4-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)Cl)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1495306
View Documents
Thermo Scientific Chemicals
A1495306
5 g
Each for $48.19
Only null left
 
AAA1495314
View Documents
Thermo Scientific Chemicals
A1495314
25 g
Each for $155.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2252-51-9
174.555
GRPWQLDSGNZEQE-UHFFFAOYSA-N
75259
C1=CC(=C(C=C1F)Cl)C(=O)O
C7H4ClFO2
MFCD00010615
benzoic acid, 2-chloro-4-fluoro, 2-chloro-4-fluoro-benzoic acid, 2-chloro-4-fluor obenzoic acid, pubchem1370, acmc-209fx5, ksc207e4n, rarechem al bo 0405, 2-chloro 4-fluoro benzoic acid, 2-chloro-4-fluoro benzoic acid, 4-fluoro-2-chloro-benzoic acid
2-chloro-4-fluorobenzoic acid

Specifications

2252-51-9
C7H4ClFO2
5 g
benzoic acid, 2-chloro-4-fluoro, 2-chloro-4-fluoro-benzoic acid, 2-chloro-4-fluor obenzoic acid, pubchem1370, acmc-209fx5, ksc207e4n, rarechem al bo 0405, 2-chloro 4-fluoro benzoic acid, 2-chloro-4-fluoro benzoic acid, 4-fluoro-2-chloro-benzoic acid
C1=CC(=C(C=C1F)Cl)C(=O)O
174.555
174.56
2-Chloro-4-fluorobenzoic acid
183°C to 187°C
MFCD00010615
1946215
GRPWQLDSGNZEQE-UHFFFAOYSA-N
2-chloro-4-fluorobenzoic acid
75259
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 218-845-0

TSCA : Yes

SDS
Documents

Documents

RUO – Research Use Only