missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-4-fluoro-5-nitrotoluene 98.0+%, TCI America™
Supplier: TCI America C327325G
Specifications
| 2-Chloro-4-fluoro-5-nitrotoluene | |
| 40°C | |
| C7H5ClFNO2 | |
| 25 g | |
| YXVJHZWHPLOEAP-UHFFFAOYSA-N | |
| 1-chloro-5-fluoro-2-methyl-4-nitrobenzene | |
| 10103979 | |
| ≥98.0% (GC) |
| 112108-73-3 | |
| Yellow | |
| MFCD11110549 | |
| 2-chloro-4-fluoro-5-nitrotoluene, 1-chloro-5-fluoro-2-methyl-4-nitro-benzene, benzene, 1-chloro-5-fluoro-2-methyl-4-nitro, acmc-2099cn, ksc682i1p, 2-chloro-4fluoro-5-nitrotoluene, 1-chloranyl-5-fluoranyl-2-methyl-4-nitro-benzene | |
| CC1=CC(=C(C=C1Cl)F)[N+](=O)[O-] | |
| 189.57 | |
| 189.57 | |
| Crystalline Powder |
Chemical Identifiers
| 112108-73-3 | |
| 189.57 | |
| YXVJHZWHPLOEAP-UHFFFAOYSA-N | |
| 10103979 | |
| CC1=CC(=C(C=C1Cl)F)[N+](=O)[O-] |
| C7H5ClFNO2 | |
| MFCD11110549 | |
| 2-chloro-4-fluoro-5-nitrotoluene, 1-chloro-5-fluoro-2-methyl-4-nitro-benzene, benzene, 1-chloro-5-fluoro-2-methyl-4-nitro, acmc-2099cn, ksc682i1p, 2-chloro-4fluoro-5-nitrotoluene, 1-chloranyl-5-fluoranyl-2-methyl-4-nitro-benzene | |
| 1-chloro-5-fluoro-2-methyl-4-nitrobenzene |
Safety and Handling
TSCA : No