missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoro-5-nitrotoluene 98.0+%, TCI America™

Catalog No. p-7125913
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
C32735G 5 g
C327325G 25 g
2 options

Catalog No. C32735G

Supplier: TCI America C32735G

Only null left

Chemical Identifiers

CAS 112108-73-3
Molecular Formula C7H5ClFNO2
Molecular Weight (g/mol) 189.57
MDL Number MFCD11110549
InChI Key YXVJHZWHPLOEAP-UHFFFAOYSA-N
Synonym 2-chloro-4-fluoro-5-nitrotoluene, 1-chloro-5-fluoro-2-methyl-4-nitro-benzene, benzene, 1-chloro-5-fluoro-2-methyl-4-nitro, acmc-2099cn, ksc682i1p, 2-chloro-4fluoro-5-nitrotoluene, 1-chloranyl-5-fluoranyl-2-methyl-4-nitro-benzene
PubChem CID 10103979
IUPAC Name 1-chloro-5-fluoro-2-methyl-4-nitrobenzene
SMILES CC1=CC(=C(C=C1Cl)F)[N+](=O)[O-]

Specifications

Melting Point 40°C
Color Yellow
Quantity 5 g
Formula Weight 189.57
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4-fluoro-5-nitrotoluene