missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoro-1-nitrobenzene, 98%

Catalog No. AAH3290603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3290603 1 g
AAH3290606 5 g
AAH3290614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3290603 Supplier Thermo Scientific Chemicals Supplier No. H3290603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2106-50-5 | C6H3ClFNO2 | 175.543 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2106-50-5
Molecular Formula C6H3ClFNO2
Molecular Weight (g/mol) 175.543
MDL Number MFCD03412200
InChI Key KQOOFMWRLDRDAX-UHFFFAOYSA-N
Synonym 2-chloro-4-fluoronitrobenzene, 1-chloro-5-fluoro-2-nitrobenzene, benzene, 2-chloro-4-fluoro-1-nitro, pubchem4342, acmc-209fh8, 2-choro-4-fluoronitrobenzene, ksc205m1p, 3-chloro-4-nitrofluorobenzene, 4-fluoro-2-chloronitrobenzene, 2-chloro-4-fluoronitrobenzenen
PubChem CID 75017
IUPAC Name 2-chloro-4-fluoro-1-nitrobenzene
SMILES C1=CC(=C(C=C1F)Cl)[N+](=O)[O-]

Specifications

Melting Point 33°C to 37°C
Odor Characteristic
Quantity 1 g
UN Number UN2811
Formula Weight 175.55
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-fluoro-1-nitrobenzene
Hazard Category H302-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only