missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,5-dimethylphenol, 98%, Thermo Scientific Chemicals

Catalog No. AAL0162706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0162706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0162706 Supplier Thermo Scientific Chemicals Supplier No. L0162706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 1124-04-5 | Molecular Formula: C8H9ClO | Molecular Weight: 156.609 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1124-04-5
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00002173
InChI Key PSOJLBXHRBFLLQ-UHFFFAOYSA-N
Synonym banol phenol, phenol, 2-chloro-4,5-dimethyl, 2-chloro-4,5-xylenol, 6-chloro-3,4-xylenol, 3,4-dimethyl-6-chlorophenol, 3,4-xylenol, 6-chloro, acmc-20anzv, 3, 6-chloro, dsstox_cid_21404, dsstox_rid_79715
PubChem CID 70754
IUPAC Name 2-chloro-4,5-dimethylphenol
SMILES CC1=CC(=C(C=C1C)Cl)O

Specifications

Melting Point 70°C to 72°C
Quantity 5 g
Beilstein 2042057
Formula Weight 156.61
Percent Purity 98%
Chemical Name or Material 2-Chloro-4,5-dimethylphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-386-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only