accessibility menu, dialog, popup

UserName

CH3OHClH3C

CAS RN 1124-04-5

IUPAC Name: 2-chloro-4,5-dimethylphenol
Synonyms: 2-chloro-4,5-dimethylphenol
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: PSOJLBXHRBFLLQ-UHFFFAOYSA-N
SMILES: CC1=CC(O)=C(Cl)C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results