missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4,5-difluorobenzoic Acid 98.0+%, TCI America™

Catalog No. C146025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C146025G 25 g
C1460250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C146025G Supplier TCI America Supplier No. C146025G

Chemical Identifiers

CAS 110877-64-0
Molecular Formula C7H2ClF2O2
Molecular Weight (g/mol) 191.54
MDL Number MFCD00077479
InChI Key CGFMLBSNHNWJAW-UHFFFAOYSA-M
Synonym 2-chloro-4,5-difluoro-benzoic acid, 2'-chloro-4',5'-difluorobenzoic acid, benzoic acid, 2-chloro-4,5-difluoro, pubchem1339, ksc177o4l, rarechem al bo 1000, timtec-bb sbb003616, 2-chloro-4,5-difluorobenzoic acid, benzoicacid, 2-chloro-4,5-difluoro
PubChem CID 737171
IUPAC Name 2-chloro-4,5-difluorobenzoate
SMILES [O-]C(=O)C1=CC(F)=C(F)C=C1Cl

Specifications

Melting Point 105°C
Color White
Quantity 25 g
Formula Weight 192.55
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-4,5-difluorobenzoic Acid
EINECSNumber (3)-4165
TSCA No