accessibility menu, dialog, popup

UserName

OOFFCl

CAS RN 110877-64-0

IUPAC Name: 2-chloro-4,5-difluorobenzoate
Synonyms: 2-chloro-4,5-difluorobenzoate
Molecular Weight (g/mol): 191.54
Molecular Formula: C7H2ClF2O2
InChi Key: CGFMLBSNHNWJAW-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(F)=C(F)C=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results