missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(4-morpholinylcarbonyl)benzeneboronic acid, 97%, Thermo Scientific™

Catalog No. p-7036885
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5534003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5534003 Supplier Thermo Scientific Chemicals Supplier No. H5534003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 2000187-73-3
Molecular Formula C11H13BClNO4
Molecular Weight (g/mol) 269.488
MDL Number MFCD22125139
InChI Key OQKJPQSIYRSBBW-UHFFFAOYSA-N
Synonym 2-chloro-4-4-morpholinylcarbonyl benzeneboronic acid, 2-chloro-4-4-morpholinylcarbonyl phenylboronic acid, 2-chloro-4-morpholine-4-carbonyl phenylboronic acid
PubChem CID 73996122
IUPAC Name [2-chloro-4-(morpholine-4-carbonyl)phenyl]boronic acid
SMILES B(C1=C(C=C(C=C1)C(=O)N2CCOCC2)Cl)(O)O

Specifications

Quantity 1 g
Formula Weight 269.49
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(4-morpholinylcarbonyl)benzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures