accessibility menu, dialog, popup

UserName

BONOClHOOH

CAS RN 2000187-73-3

IUPAC Name: [2-chloro-4-(morpholine-4-carbonyl)phenyl]boronic acid
Synonyms: 2-chloro-4-(morpholine-4-carbonyl)phenylboronic acid
Molecular Weight (g/mol): 269.49
Molecular Formula: C11H13BClNO4
InChi Key: OQKJPQSIYRSBBW-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C=C(C=C1)C(=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results