missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-3-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. C23335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23335G 5 g
C233325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23335G Supplier TCI America Supplier No. C23335G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3970-35-2
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
MDL Number MFCD00007069
InChI Key JRQDVRIQJJPHEQ-UHFFFAOYSA-N
Synonym 2-chloro-3-nitrobenzoic acid, 2-chloro-3-nitro-benzoic acid, 2-chloro-3-nitrobenzoicacid, benzoic acid, 2-chloro-3-nitro, 2-chloro-3-carboxynitrobenzene, nitrochlorbenzoesaure, pubchem2100, acmc-1ctvl, 2-chloro-3-nitrobenzic acid, 2-chloro-3-nitrobezoic acid
PubChem CID 19870
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])Cl)C(=O)O

Specifications

Melting Point 185°C
Color White-Yellow
Quantity 5 g
Formula Weight 201.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-3-nitrobenzoic Acid
EINECSNumber (3)-1502
TSCA No