accessibility menu, dialog, popup

UserName

OHOClNOO

CAS RN 3970-35-2

IUPAC Name: 2-chloro-3-nitrobenzoic acid
Synonyms: 2-chloro-3-nitrobenzoic acid
Molecular Weight (g/mol): 201.56
Molecular Formula: C7H4ClNO4
InChi Key: JRQDVRIQJJPHEQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1Cl)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)