missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-2-phenylacetophenone 98.0+%, TCI America™
Supplier: TCI America D174525G
Specifications
| 2-Chloro-2-phenylacetophenone | |
| 68°C | |
| C14H11ClO | |
| 25 g | |
| desyl chloride, 2-chloro-2-phenylacetophenone, alpha-chlorodeoxybenzoin, 1,2-diphenyl-2-chloroethanone, ethanone, 2-chloro-1,2-diphenyl, alpha-chlorobenzyl phenyl ketone, 2-chloro-1,2-diphenylethone, acetophenone, alpha-chloro-alpha-phenyl, 2-chloro-1,2-diphenyl-ethanone, acetophenone, 2-chloro-2-phenyl | |
| ClC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 230.69 | |
| 230.69 | |
| Crystalline Powder |
| 447-31-4 | |
| White-Yellow | |
| MFCD00000858 | |
| 3261 | |
| RXDYOLRABMJTEF-UHFFFAOYNA-N | |
| 2-chloro-1,2-diphenylethan-1-one | |
| 95343 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 447-31-4 | |
| 230.69 | |
| RXDYOLRABMJTEF-UHFFFAOYNA-N | |
| 95343 | |
| ClC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1 |
| C14H11ClO | |
| MFCD00000858 | |
| desyl chloride, 2-chloro-2-phenylacetophenone, alpha-chlorodeoxybenzoin, 1,2-diphenyl-2-chloroethanone, ethanone, 2-chloro-1,2-diphenyl, alpha-chlorobenzyl phenyl ketone, 2-chloro-1,2-diphenylethone, acetophenone, alpha-chloro-alpha-phenyl, 2-chloro-1,2-diphenyl-ethanone, acetophenone, 2-chloro-2-phenyl | |
| 2-chloro-1,2-diphenylethan-1-one |
Safety and Handling
RTECSNumber : AM6825000
TSCA : Yes