accessibility menu, dialog, popup

UserName

ClO

CAS RN 447-31-4

IUPAC Name: 2-chloro-1,2-diphenylethan-1-one
Synonyms: 2-chloro-1,2-diphenylethanone
Molecular Weight (g/mol): 230.69
Molecular Formula: C14H11ClO
InChi Key: RXDYOLRABMJTEF-UHFFFAOYNA-N
SMILES: ClC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)