missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-2-phenylacetophenone 98.0+%, TCI America™

Catalog No. p-7126908
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17455G 5 g
D174525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17455G Supplier TCI America Supplier No. D17455G
Only null left

Chemical Identifiers

CAS 447-31-4
Molecular Formula C14H11ClO
Molecular Weight (g/mol) 230.69
MDL Number MFCD00000858
InChI Key RXDYOLRABMJTEF-UHFFFAOYNA-N
Synonym desyl chloride, 2-chloro-2-phenylacetophenone, alpha-chlorodeoxybenzoin, 1,2-diphenyl-2-chloroethanone, ethanone, 2-chloro-1,2-diphenyl, alpha-chlorobenzyl phenyl ketone, 2-chloro-1,2-diphenylethone, acetophenone, alpha-chloro-alpha-phenyl, 2-chloro-1,2-diphenyl-ethanone, acetophenone, 2-chloro-2-phenyl
PubChem CID 95343
IUPAC Name 2-chloro-1,2-diphenylethan-1-one
SMILES ClC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 230.69
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-2-phenylacetophenone
RTECSNumber AM6825000
TSCA Yes