missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1-methylpyridinium p-Toluenesulfonate 98.0+%, TCI America™

Catalog No. c090625g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C090625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C090625G Supplier TCI America Supplier No. C090625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7403-46-5
Molecular Formula C13H14ClNO3S
Molecular Weight (g/mol) 299.769
MDL Number MFCD00011985
InChI Key KWNGIKVZXFFZNN-UHFFFAOYSA-M
PubChem CID 10990220
IUPAC Name 2-chloro-1-methylpyridin-1-ium;4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)[O-].C[N+]1=CC=CC=C1Cl

Specifications

Melting Point 125°C
Color White-Yellow
Quantity 25 g
Formula Weight 299.77
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloro-1-methylpyridinium p-Toluenesulfonate
TSCA No
Recommended Storage Refrigerator