accessibility menu, dialog, popup

UserName

CAS RN 7403-46-5

IUPAC Name: 2-chloro-1-methylpyridin-1-ium 4-methylbenzene-1-sulfonate
Synonyms: 2-chloro-1-methylpyridin-1-ium tosylate
Molecular Weight (g/mol): 299.77
Molecular Formula: C13H14ClNO3S
InChi Key: KWNGIKVZXFFZNN-UHFFFAOYSA-M
SMILES: C[N+]1=CC=CC=C1Cl.CC1=CC=C(C=C1)S([O-])(=O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results