missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-1,4-phenylenediamine Sulfate 98.0+%, TCI America™
$143.54 - $1310.43
Chemical Identifiers
| CAS | 61702-44-1 |
|---|---|
| Molecular Formula | C6H9ClN2O4S |
| Molecular Weight (g/mol) | 240.66 |
| MDL Number | MFCD00013004 |
| InChI Key | GQFGHCRXPLROOF-UHFFFAOYSA-N |
| Synonym | 2-chloro-p-phenylenediamine sulfate, 2-chlorobenzene-1,4-diamine sulfate, 2-chloro-1,4-phenylenediamine sulfate, fourrine so, 2-chloro-1,4-benzenediamine sulfate, 2-chlorobenzene-1,4-diamine; sulfuric acid, 1,4-benzenediamine, 2-chloro-, sulfate, renal so, fourrine 81, 2-cl-p-pd |
| PubChem CID | 22584 |
| ChEBI | CHEBI:76597 |
| IUPAC Name | 2-chlorobenzene-1,4-diamine; sulfuric acid |
| SMILES | OS(O)(=O)=O.NC1=CC=C(N)C(Cl)=C1 |
Chemical Identifiers
| 61702-44-1 | |
| 240.66 | |
| GQFGHCRXPLROOF-UHFFFAOYSA-N | |
| 22584 | |
| 2-chlorobenzene-1,4-diamine; sulfuric acid |
| C6H9ClN2O4S | |
| MFCD00013004 | |
| 2-chloro-p-phenylenediamine sulfate, 2-chlorobenzene-1,4-diamine sulfate, 2-chloro-1,4-phenylenediamine sulfate, fourrine so, 2-chloro-1,4-benzenediamine sulfate, 2-chlorobenzene-1,4-diamine; sulfuric acid, 1,4-benzenediamine, 2-chloro-, sulfate, renal so, fourrine 81, 2-cl-p-pd | |
| CHEBI:76597 | |
| OS(O)(=O)=O.NC1=CC=C(N)C(Cl)=C1 |
Specifications
| 61702-44-1 | |
| White-Yellow | |
| MFCD00013004 | |
| 2-chloro-p-phenylenediamine sulfate, 2-chlorobenzene-1,4-diamine sulfate, 2-chloro-1,4-phenylenediamine sulfate, fourrine so, 2-chloro-1,4-benzenediamine sulfate, 2-chlorobenzene-1,4-diamine; sulfuric acid, 1,4-benzenediamine, 2-chloro-, sulfate, renal so, fourrine 81, 2-cl-p-pd | |
| OS(O)(=O)=O.NC1=CC=C(N)C(Cl)=C1 | |
| 240.66 | |
| CHEBI:76597 | |
| ≥98.0% (HPLC,N) | |
| 2-Chloro-1,4-phenylenediamine Sulfate |
| 253°C | |
| C6H9ClN2O4S | |
| 25 g | |
| GQFGHCRXPLROOF-UHFFFAOYSA-N | |
| 2-chlorobenzene-1,4-diamine; sulfuric acid | |
| 22584 | |
| 240.66 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : CZ1581000
TSCA : No