accessibility menu, dialog, popup

UserName

NH2NH2ClOHSOHOO

CAS RN 61702-44-1

IUPAC Name: 2-chlorobenzene-1,4-diamine; sulfuric acid
Synonyms: 2-chloro-P-phenylenediamine
Molecular Weight (g/mol): 240.66
Molecular Formula: C6H9ClN2O4S
InChi Key: GQFGHCRXPLROOF-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=CC=C(N)C(Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results