missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-1,4-phenylenediamine Sulfate 98.0+%, TCI America™
Supplier: TCI America C0256500G
Specifications
| 2-Chloro-1,4-phenylenediamine Sulfate | |
| 253°C | |
| C6H9ClN2O4S | |
| 500 g | |
| GQFGHCRXPLROOF-UHFFFAOYSA-N | |
| 2-chlorobenzene-1,4-diamine; sulfuric acid | |
| 22584 | |
| 240.66 | |
| Crystalline Powder |
| 61702-44-1 | |
| White-Yellow | |
| MFCD00013004 | |
| 2-chloro-p-phenylenediamine sulfate, 2-chlorobenzene-1,4-diamine sulfate, 2-chloro-1,4-phenylenediamine sulfate, fourrine so, 2-chloro-1,4-benzenediamine sulfate, 2-chlorobenzene-1,4-diamine; sulfuric acid, 1,4-benzenediamine, 2-chloro-, sulfate, renal so, fourrine 81, 2-cl-p-pd | |
| OS(O)(=O)=O.NC1=CC=C(N)C(Cl)=C1 | |
| 240.66 | |
| CHEBI:76597 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 61702-44-1 | |
| 240.66 | |
| GQFGHCRXPLROOF-UHFFFAOYSA-N | |
| 22584 | |
| 2-chlorobenzene-1,4-diamine; sulfuric acid |
| C6H9ClN2O4S | |
| MFCD00013004 | |
| 2-chloro-p-phenylenediamine sulfate, 2-chlorobenzene-1,4-diamine sulfate, 2-chloro-1,4-phenylenediamine sulfate, fourrine so, 2-chloro-1,4-benzenediamine sulfate, 2-chlorobenzene-1,4-diamine; sulfuric acid, 1,4-benzenediamine, 2-chloro-, sulfate, renal so, fourrine 81, 2-cl-p-pd | |
| CHEBI:76597 | |
| OS(O)(=O)=O.NC1=CC=C(N)C(Cl)=C1 |
Safety and Handling
RTECSNumber : CZ1581000
TSCA : No