Learn More
2-Chloro-1,3-dinitrobenzene, 98+%
CAS: 606-21-3 | C6H3ClN2O4 | 202.55 g/mol
Supplier: Thermo Scientific Chemicals L0459306
Description
2-Chloro-1,3-dinitrobenzene can be used as intermediates of dyes and pigments. Also used as solvents in HPLC, NMR.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloro-1,3-dinitrobenzene can be used as intermediates of dyes and pigments. Also used as solvents in HPLC, NMR.
Solubility
Soluble in alcohol. Moderately soluble in benzene, ether. Practically insoluble in water.
Notes
Incompatible materials are oxidizing agents, Strong bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
| 2-Chloro-1,3-dinitrobenzene | |
| 83°C to 88°C | |
| 315°C | |
| MFCD00014689 | |
| UN3441 | |
| 14,2137 | |
| Soluble in alcohol. Moderately soluble in benzene,ether. Practically insoluble in water. | |
| [O-][N+](=O)C1=CC=CC(=C1Cl)[N+]([O-])=O | |
| 202.55 | |
| 202.55 |
| 606-21-3 | |
| 1.687 | |
| C6H3ClN2O4 | |
| 5 g | |
| 522187 | |
| 2,6-dinitrochlorobenzene, 1-chloro-2,6-dinitrobenzene, benzene, 2-chloro-1,3-dinitro, 2-chloro-1,3-dintrobenzene, 1,3-dinitro-2-chlorobenzene, 2-chloro-1,3-dinitro-benzene, unii-lg72rnd0ab, lg72rnd0ab, 1-chloro-2,3-dinitrobenzene mixture, ambsclk-136 | |
| BPPMIQPXQVIZNJ-UHFFFAOYSA-N | |
| 2-chloro-1,3-dinitrobenzene | |
| 11814 | |
| ≥98% |
Chemical Identifiers
| 606-21-3 | |
| 202.55 | |
| BPPMIQPXQVIZNJ-UHFFFAOYSA-N | |
| 11814 | |
| [O-][N+](=O)C1=CC=CC(=C1Cl)[N+]([O-])=O |
| C6H3ClN2O4 | |
| MFCD00014689 | |
| 2,6-dinitrochlorobenzene, 1-chloro-2,6-dinitrobenzene, benzene, 2-chloro-1,3-dinitro, 2-chloro-1,3-dintrobenzene, 1,3-dinitro-2-chlorobenzene, 2-chloro-1,3-dinitro-benzene, unii-lg72rnd0ab, lg72rnd0ab, 1-chloro-2,3-dinitrobenzene mixture, ambsclk-136 | |
| 2-chloro-1,3-dinitrobenzene |
Safety and Handling
GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P501c
H301+H311-H330-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLORODINITROBENZENES, SOLID
EINECSNumber : 210-107-6
RTECSNumber : CZ0525500
TSCA : Yes
RUO – Research Use Only