missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,3-dinitrobenzene, 98+%

Catalog No. p-7044103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0459303 1 g
AAL0459306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0459303 Supplier Thermo Scientific Chemicals Supplier No. L0459303

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 606-21-3 | C6H3ClN2O4 | 202.55 g/mol

2-Chloro-1,3-dinitrobenzene can be used as intermediates of dyes and pigments. Also used as solvents in HPLC, NMR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-1,3-dinitrobenzene can be used as intermediates of dyes and pigments. Also used as solvents in HPLC, NMR.

Solubility
Soluble in alcohol. Moderately soluble in benzene, ether. Practically insoluble in water.

Notes
Incompatible materials are oxidizing agents, Strong bases. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 606-21-3
Molecular Formula C6H3ClN2O4
Molecular Weight (g/mol) 202.55
MDL Number MFCD00014689
InChI Key BPPMIQPXQVIZNJ-UHFFFAOYSA-N
Synonym 2,6-dinitrochlorobenzene, 1-chloro-2,6-dinitrobenzene, benzene, 2-chloro-1,3-dinitro, 2-chloro-1,3-dintrobenzene, 1,3-dinitro-2-chlorobenzene, 2-chloro-1,3-dinitro-benzene, unii-lg72rnd0ab, lg72rnd0ab, 1-chloro-2,3-dinitrobenzene mixture, ambsclk-136
PubChem CID 11814
IUPAC Name 2-chloro-1,3-dinitrobenzene
SMILES [O-][N+](=O)C1=CC=CC(=C1Cl)[N+]([O-])=O

Specifications

Melting Point 83°C to 88°C
Density 1.687
Boiling Point 315°C
Quantity 1 g
UN Number UN3441
Beilstein 522187
Merck Index 14,2137
Solubility Information Soluble in alcohol. Moderately soluble in benzene,ether. Practically insoluble in water.
Formula Weight 202.55
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-1,3-dinitrobenzene
Show More Show Less
Hazard Category H301+H311-H330-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLORODINITROBENZENES, SOLID
EINECSNumber 210-107-6
RTECSNumber CZ0525500
TSCA Yes

RUO – Research Use Only