missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Carboxyphenyl Salicylate 98.0+%, TCI America™
Supplier: TCI America S012925G
Specifications
| 2-Carboxyphenyl Salicylate | |
| 137°C | |
| C14H10O5 | |
| 25 g | |
| WVYADZUPLLSGPU-UHFFFAOYSA-N | |
| 2-(2-hydroxybenzoyl)oxybenzoic acid | |
| 5161 | |
| 258.23 | |
| Crystalline Powder |
| 552-94-3 | |
| White | |
| MFCD00020252 | |
| salsalate, sasapyrine, salicylsalicylic acid, disalicylic acid, disalcid, sasapyrinum, saloxium, 2-carboxyphenyl salicylate, salina, diacesal | |
| C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O | |
| 258.229 | |
| CHEBI:9014 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 552-94-3 | |
| 258.229 | |
| WVYADZUPLLSGPU-UHFFFAOYSA-N | |
| 5161 | |
| 2-(2-hydroxybenzoyl)oxybenzoic acid |
| C14H10O5 | |
| MFCD00020252 | |
| salsalate, sasapyrine, salicylsalicylic acid, disalicylic acid, disalcid, sasapyrinum, saloxium, 2-carboxyphenyl salicylate, salina, diacesal | |
| CHEBI:9014 | |
| C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O |
Safety and Handling
EINECSNumber : (3)-1534
RTECSNumber : DH2080000
TSCA : Yes