accessibility menu, dialog, popup

UserName

CAS RN 552-94-3

IUPAC Name: 2-(2-hydroxybenzoyloxy)benzoic acid
Synonyms: salina 3M brand of salicylsalicylic acid diacesal salicyloylsalicylic acid sasapyrinum sasapyrine diplosal disalicylic acid argesic salflex
Molecular Weight (g/mol): 258.23
Molecular Formula: C14H10O5
InChi Key: WVYADZUPLLSGPU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1OC(=O)C1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)