missing translation for 'onlineSavingsMsg'
Learn More

2-Carboxyphenyl Salicylate 98.0+%, TCI America™

$54.06 - $54.06

Chemical Identifiers

CAS 552-94-3
Molecular Formula C14H10O5
Molecular Weight (g/mol) 258.229
MDL Number MFCD00020252
InChI Key WVYADZUPLLSGPU-UHFFFAOYSA-N
Synonym salsalate, sasapyrine, salicylsalicylic acid, disalicylic acid, disalcid, sasapyrinum, saloxium, 2-carboxyphenyl salicylate, salina, diacesal
PubChem CID 5161
ChEBI CHEBI:9014
IUPAC Name 2-(2-hydroxybenzoyl)oxybenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S012925G
View Documents
TCI America
S012925G
25 g
Each for $54.06
Only null left
 
Description

Description

Specifications

Chemical Identifiers

552-94-3
258.229
WVYADZUPLLSGPU-UHFFFAOYSA-N
5161
2-(2-hydroxybenzoyl)oxybenzoic acid
C14H10O5
MFCD00020252
salsalate, sasapyrine, salicylsalicylic acid, disalicylic acid, disalcid, sasapyrinum, saloxium, 2-carboxyphenyl salicylate, salina, diacesal
CHEBI:9014
C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O

Specifications

552-94-3
White
MFCD00020252
salsalate, sasapyrine, salicylsalicylic acid, disalicylic acid, disalcid, sasapyrinum, saloxium, 2-carboxyphenyl salicylate, salina, diacesal
C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O
258.229
CHEBI:9014
≥98.0% (HPLC,T)
2-Carboxyphenyl Salicylate
137°C
C14H10O5
25 g
WVYADZUPLLSGPU-UHFFFAOYSA-N
2-(2-hydroxybenzoyl)oxybenzoic acid
5161
258.23
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1534

RTECSNumber : DH2080000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'