missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Carboxyethyl)phenylphosphinic Acid 98.0+%, TCI America™
$37.29 - $132.35
Chemical Identifiers
| CAS | 14657-64-8 |
|---|---|
| Molecular Formula | C9H11O4P |
| Molecular Weight (g/mol) | 214.157 |
| MDL Number | MFCD08704562 |
| InChI Key | MORLYCDUFHDZKO-UHFFFAOYSA-N |
| PubChem CID | 11287340 |
| IUPAC Name | 3-[hydroxy(phenyl)phosphoryl]propanoic acid |
| SMILES | C1=CC=C(C=C1)P(=O)(CCC(=O)O)O |
Chemical Identifiers
| 14657-64-8 | |
| 214.157 | |
| MORLYCDUFHDZKO-UHFFFAOYSA-N | |
| 3-[hydroxy(phenyl)phosphoryl]propanoic acid |
| C9H11O4P | |
| MFCD08704562 | |
| 11287340 | |
| C1=CC=C(C=C1)P(=O)(CCC(=O)O)O |
Specifications
| 14657-64-8 | |
| White | |
| MFCD08704562 | |
| MORLYCDUFHDZKO-UHFFFAOYSA-N | |
| 3-[hydroxy(phenyl)phosphoryl]propanoic acid | |
| 11287340 | |
| ≥98.0% (HPLC,T) | |
| (2-Carboxyethyl)phenylphosphinic Acid |
| 160°C | |
| C9H11O4P | |
| 5 g | |
| C1=CC=C(C=C1)P(=O)(CCC(=O)O)O | |
| 214.157 | |
| 214.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No