accessibility menu, dialog, popup

UserName

POOOHHO

CAS RN 14657-64-8

IUPAC Name: 3-[hydroxy(phenyl)phosphoryl]propanoic acid
Synonyms: 3-[hydroxy(phenyl)phosphoryl]propanoic acid
Molecular Weight (g/mol): 214.16
Molecular Formula: C9H11O4P
InChi Key: MORLYCDUFHDZKO-UHFFFAOYNA-N
SMILES: OC(=O)CCP(O)(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results