missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Carboxyethyl)phenylphosphinic Acid 98.0+%, TCI America™
Supplier: TCI America C288725G
Specifications
| (2-Carboxyethyl)phenylphosphinic Acid | |
| 160°C | |
| C9H11O4P | |
| 25 g | |
| C1=CC=C(C=C1)P(=O)(CCC(=O)O)O | |
| 214.157 | |
| 214.16 | |
| Crystalline Powder |
| 14657-64-8 | |
| White | |
| MFCD08704562 | |
| MORLYCDUFHDZKO-UHFFFAOYSA-N | |
| 3-[hydroxy(phenyl)phosphoryl]propanoic acid | |
| 11287340 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 14657-64-8 | |
| 214.157 | |
| MORLYCDUFHDZKO-UHFFFAOYSA-N | |
| 3-[hydroxy(phenyl)phosphoryl]propanoic acid |
| C9H11O4P | |
| MFCD08704562 | |
| 11287340 | |
| C1=CC=C(C=C1)P(=O)(CCC(=O)O)O |
Safety and Handling
TSCA : No